Cu Fe Al Mn Sb Mo Ag Ni As Pb Ba Be Sn B Tl Cd U Co V Cr Zn in HNO3 2% 1 * 100 mL
Catalog # CMPLRM103297L1
Supplier: CUSTOM MADE CHEMICALS LAB
Cu Fe Al Mn Sb Mo Ag Ni As Pb Ba Be Sn B Tl Cd U Co V Cr Zn in HNO3 2% 1 * 100 mL
Catalog # CMPLRM103297L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test strips, nitrite, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Test kits, chloride, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Accessories
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach