ContactUsLinkComponent
The MassivEV™ EV Purification Column PS and MassivEV™ Purification Buffer Set have been specifically designed for the isolation and purification of extracellular vesicles (EVs), including exosomes, from large volumes of cell culture supernatants. Thi 1 * 1 mL
Catalog # WAKF131-19491
The MassivEV™ EV Purification Column PS and MassivEV™ Purification Buffer Set have been specifically designed for the isolation and purification of extracellular vesicles (EVs), including exosomes, from large volumes of cell culture supernatants. Thi 1 * 1 mL
Catalog # WAKF131-19491
Frequently Bought Together