ContactUsLinkComponent
Calcium 10 g/l in diluted HNO3 for ICP 1 * 100 mL
Catalog # ACSDC109.2NP.L1
Supplier:  CPACHEM
Calcium 10 g/l in diluted HNO3 for ICP 1 * 100 mL
Catalog # ACSDC109.2NP.L1
Supplier:  CPACHEM
Frequently Bought Together