ContactUsLinkComponent
Bacteriophage T7 RNA Polymerase is a DNA-dependent RNA polymerase with high specificity for the T7 promoter. This enzyme catalyzes t he 5Æ to 3Æ synthesis of RNA from DNA downstream from its promoter.This product with activity data is belong to b 1 * 25.000 Unit
Catalog # ABNOP8870.25000
Supplier:  Abnova
Bacteriophage T7 RNA Polymerase is a DNA-dependent RNA polymerase with high specificity for the T7 promoter. This enzyme catalyzes t he 5Æ to 3Æ synthesis of RNA from DNA downstream from its promoter.This product with activity data is belong to b 1 * 25.000 Unit
Catalog # ABNOP8870.25000
Supplier:  Abnova
Frequently Bought Together