Labeled FISH probes for identification of gene split using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
ABNOFS0100.100UL
: Abnova
Labeled FISH probes for identification of gene split using Fluorescent In Situ Hybridization Technique. (Technology) 1 * 100 µl
ABNOFS0100.100UL
: Abnova
Frequently Bought Together
VWR®, Cryovials
: VWR Collection
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
Screw caps for laboratory/media bottles
: SciLabware
MicroWell™ Plates, 96 well, Nunc™
: Thermo Fisher Scientific
Microcentrifuge tubes, 2,0 ml
: Brand
VWR®, Sample Dippers
: VWR Collection
Cryogenic tubes, CryoTubes™, Nunc™
: Thermo Fisher Scientific
Cell culture plates, Nunc™ Multidish
: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Microcentrifuge tubes, 0,6/1,5/2,0 ml, graduated, with snap cap
: Thermo Fisher Scientific
Storage tubes, 2D barcoded, Matrix™
: THERMO MATRIX TECHNOLOGIES
VWR®, Blotting Pads, Grade 707
: VWR Collection
Petri dishes
: Anicrin
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
VWR® Cell Chambers
: VWR Collection
Stoppers for tubes
: Greiner Bio-One