300 ML. 8 RESERVOIR BASES. NON-STERILE. 1 * 8 items
Catalog # INMS6305
Supplier: INTEGRA
Frequently Bought Together
HINGE SLEEVES (PLASTIC) 1 * 10 ST
Supplier: PLASTIC HAUS
Bags, zip seal, anti-UV, writing area, Minigrip®
Supplier: Flexico
Recombinant human CD4 protein Active 1 * 50 µG
Supplier: Abcam
NIERENSCHALE 700 ML STERILE AND BLAUE 1 * 60 ST
Supplier: MARKET SOURCE PART PROCESS
[EN]WEBER GARBAGE BAGS 35 LT GRAY 1 * 25 Rolle
Supplier: MARKET SOURCE PART PROCESS
Pressure caps for jars
Supplier: LAMAPLAST
STORAGE BIN SSI SCHÄFER LF 321, POLYPROPYLENE, L 343 X W 209 X H 145 MM, 7.5 L, BLUE 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Bottles, wide neck, square with thread
Supplier: MAUSER
Connection system GL 45
Supplier: SciLabware