5-(DIFLUOROMETHYL)-1-(1-{[(9H-FLUOREN-9-YL)METHOXY]CARBONYL}PIPERIDIN-4-YL)-1H-1,2,3-TRIAZOLE-4-CARBOXYLIC ACID 1 * 1 g
Catalog # UKROCSSB0010267998
Supplier: UKRORGSYNTHESIS
5-(DIFLUOROMETHYL)-1-(1-{[(9H-FLUOREN-9-YL)METHOXY]CARBONYL}PIPERIDIN-4-YL)-1H-1,2,3-TRIAZOLE-4-CARBOXYLIC ACID 1 * 1 g
Catalog # UKROCSSB0010267998
Supplier: UKRORGSYNTHESIS
Frequently Bought Together
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Diethyl carbonate for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide, Bioreagent
Supplier: Thermo Fisher Scientific
Pyridine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Acetone, Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Heptafluorobutyric anhydride (HFAA), Supelco®
Supplier: Merck
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% for HPLC
Supplier: VWR Chemicals