ContactUsLinkComponent
GAM Broth modified is used for the isolation and cultivation of anaerobic bacteria from clinical samples. It is also used for antibiotic sensitivity tests (other than sulphonamide drugs). 1 * 1 items
Catalog # NISS05433-GBM-0100
GAM Broth modified is used for the isolation and cultivation of anaerobic bacteria from clinical samples. It is also used for antibiotic sensitivity tests (other than sulphonamide drugs). 1 * 1 items
Catalog # NISS05433-GBM-0100
Frequently Bought Together