Orders LinkContactUsLinkComponent
Bosshead
Bosshead
Thumb screws, 13 mm, Aluminium alloy, powder coated
Catalog # USBE2420
Supplier:   USBECK Laborgeräte
Bosshead
Catalog # USBE2420
Bosshead
Bosshead
Thumb screws, 13 mm, Aluminium alloy, powder coated
Price
SFr. 22.80
Log in for availability
1 * 1 ST
Product Details & Documents
For rods up to 13 mm. Adjustable, nickel plated brass screws.
Specifications
  • Spanning width:
    13 mm
  • Description:
    Thumb screws
  • Material:
    Aluminium alloy, powder coated