Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Cerex® TRACE-B SPE Columns

Column Cerex TRACE-B, 15 µm, 3 ml, 35 mg
  TCAT711-335C
 :   Tecan
Cerex® TRACE-B SPE Columns
  TCAT711-335C
 :  Tecan
Cerex® TRACE-B SPE Columns

Column Cerex TRACE-B, 15 µm, 3 ml, 35 mg

 
 
   
1 * 100 ST
 

Strong cationic exchange chemistry ideal for aggressive cleanup of basic target compounds.

  • Ion-exchange
  • Liquid handler compatible

A 15 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.

 
  • Particle Size:
    15 µm
  • Capacity:
    3 ml
  • Description:
    Column Cerex TRACE-B
  • Seq:
    03
  • Bed weight:
    35 mg