ContactUsLinkComponent
8 components: 1000mg/l each of N-ISOPROPYL-N'-PHENYL-1 ,4-PHENYLENEDIAMINE [CAS:101-72-4] N-(1,3-Dimethylbutyl)-N'-phenyl-1 ,4-phenylenediamine [CAS:793-24-8] 2,6-Di-tert-butyl-4-methylphenol [CAS:128-37-0] Pentaerythritol Tetrakis [CAS:6683-19 1 * 1 mL
Catalog # CMPLRD0986191
8 components: 1000mg/l each of N-ISOPROPYL-N'-PHENYL-1 ,4-PHENYLENEDIAMINE [CAS:101-72-4] N-(1,3-Dimethylbutyl)-N'-phenyl-1 ,4-phenylenediamine [CAS:793-24-8] 2,6-Di-tert-butyl-4-methylphenol [CAS:128-37-0] Pentaerythritol Tetrakis [CAS:6683-19 1 * 1 mL
Catalog # CMPLRD0986191
Frequently Bought Together