ContactUsLinkComponent
13 components: S 10000mg/l Mg 2000mg/l Na 1000mg/l Fe 100mg/l Mn 100mg/l Zn 100mg/l Cr 1mg/l Li 1mg/l Ni 1mg/l V 1mg/l Be 0.5mg/l Cd 0.5mg/l Co 0.5mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 8 months. (4 months a 1 * 5 Ampoul
Catalog # CMPLRM082332L1.5
13 components: S 10000mg/l Mg 2000mg/l Na 1000mg/l Fe 100mg/l Mn 100mg/l Zn 100mg/l Cr 1mg/l Li 1mg/l Ni 1mg/l V 1mg/l Be 0.5mg/l Cd 0.5mg/l Co 0.5mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 8 months. (4 months a 1 * 5 Ampoul
Catalog # CMPLRM082332L1.5
Frequently Bought Together