Citrate - Disodium Hydrogen Phosphate Buffer Solution (pH 7.0) 1 * 1 L
ACSDCH8004017.1L
: CPACHEM
Frequently Bought Together
Acetonitrile, anhydrous ≥99.8%
: Thermo Fisher Scientific
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
: Thermo Fisher Scientific
Acetonitrile LC-MS 2.5L 1 * 1 ST
: MARKET SOURCE PART PROCESS
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals