ugo basile Elevated Zero Maze for Mouse, with non-reflecting grey external walls, internal IR walls, 3mm edge on open part. Floor w ith non-smooth texture and high contrast grey painted. 1 * 1 ST
Catalog # MSPP40165
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together