ContactUsLinkComponent
Grinding jar EasyFit for Planetary Ball Mills, agate, 80ml 1 * 1 items
Catalog # RETS01.462.0511
Supplier:  Retsch GmbH
Grinding jar EasyFit for Planetary Ball Mills, agate, 80ml 1 * 1 items
Catalog # RETS01.462.0511
Supplier:  Retsch GmbH
Frequently Bought Together