ContactUsLinkComponent
Kit: Barcode label thermal printer (SRvooo-0484) and 2 paper rolls 57 mm (THE002-011B) used for cycle documentation. During the cycle the printer records temperature, pressure, date, cycle status, end of cycle. 1 * 1 KIT
Catalog # TUTTSRV000-0485
Supplier:  TUTTNAUER
Kit: Barcode label thermal printer (SRvooo-0484) and 2 paper rolls 57 mm (THE002-011B) used for cycle documentation. During the cycle the printer records temperature, pressure, date, cycle status, end of cycle. 1 * 1 KIT
Catalog # TUTTSRV000-0485
Supplier:  TUTTNAUER
Frequently Bought Together