ICP STANDARD 27 COMPONENTS 27 components: Ag 10mg/l Al 10mg/l As 1000mg/l B 10mg/l Ba 10mg/l Ca 100mg/l Cd 10mg/l Co 10mg/l Cr 10mg/l Cu 10mg/l Fe 100mg/l K 100mg/l Li 10mg/l Mg 10mg/l Mn 10mg/l Mo 10mg/l Na 100mg/l 1 * 1 items
Catalog # CMPLRM044259L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together