COPPER(I) OXIDE 99.5% - 500G 1 * 500 g
Catalog # WAKO033-13205
Supplier: Wako
Frequently Bought Together
Barium perchlorate, EMSURE® for analysis, Supelco®
Supplier: Merck
Parteck® MST (Magnesium stearate), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, FCC vegetable grade, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Calcium sulphate, Drierite™ 6 mm (4 mesh) for drying, with indicator (cobalt (II) chloride)
Supplier: Thermo Fisher Scientific
Zinc stearate, powder USP
Supplier: Spectrum Chemical
Disodium hydrogen phosphate dodecahydrate ≥98%, purified
Supplier: VWR Chemicals
Nickel(II) nitrate hexahydrate 99.995%
Supplier: Apollo Scientific
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals