ContactUsLinkComponent
5 ampoules de 1 mL ,,13 components: 100mg/l each of Paracetamol [CAS:103-90-2] Caffeine [CAS:58-08-2] 1H-Benzotriazole [CAS:95-14-7] Tolyltriazole [CAS:29385-43-1] Bisphenol S [CAS:80-09-1] 4-Hydroxybenzoic acid-methyl ester [CAS:99-76-3] 1 * 5 Ampoul
Catalog # CMPLRD0990281.5
5 ampoules de 1 mL ,,13 components: 100mg/l each of Paracetamol [CAS:103-90-2] Caffeine [CAS:58-08-2] 1H-Benzotriazole [CAS:95-14-7] Tolyltriazole [CAS:29385-43-1] Bisphenol S [CAS:80-09-1] 4-Hydroxybenzoic acid-methyl ester [CAS:99-76-3] 1 * 5 Ampoul
Catalog # CMPLRD0990281.5
Frequently Bought Together