LABNETWORK 4-BROMO-5-METHYLBENZENE-1,3-DIOL 1 * 25 g
Catalog # MSPPWXCD0063317925
Supplier: MARKET SOURCE PART PROCESS
LABNETWORK 4-BROMO-5-METHYLBENZENE-1,3-DIOL 1 * 25 g
Catalog # MSPPWXCD0063317925
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Paraffin, liquid, liquid Ph. Eur.
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals