GRAPE FLAVOR ARTIFICIAL CONCENTRATE 1 * 25 mL
Catalog # SPCMG1427-25ML
Supplier: Spectrum Chemical
Frequently Bought Together
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 80…100 °C, EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% for peptide synthesis
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine 99+%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals