ContactUsLinkComponent
8 components: Dextran 40 [CAS:9004-54-0] 2g/l D-(+)-Maltose Monohydrate [CAS:6363-53-7] 2.4g/l Maltotriose [CAS:1109-28-0] 2.4g/l D-(+)-Glucose [CAS:50-99-7] 4g/l L-(+)-Lactic acid [CAS:79-33-4] 2g/l Acetic acid [CAS:64-19-7] 0.4g/l Ethan 1 * 10 items
Catalog # CMPLRD10003110.10
8 components: Dextran 40 [CAS:9004-54-0] 2g/l D-(+)-Maltose Monohydrate [CAS:6363-53-7] 2.4g/l Maltotriose [CAS:1109-28-0] 2.4g/l D-(+)-Glucose [CAS:50-99-7] 4g/l L-(+)-Lactic acid [CAS:79-33-4] 2g/l Acetic acid [CAS:64-19-7] 0.4g/l Ethan 1 * 10 items
Catalog # CMPLRD10003110.10
Frequently Bought Together