Stablcal Verification Vial, 1 NTU, without RFID, TU5200 1 * 1 items
Catalog # HACHLZZ004
Supplier: Hach
Stablcal Verification Vial, 1 NTU, without RFID, TU5200 1 * 1 items
Catalog # HACHLZZ004
Supplier: Hach
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH combination electrodes, IntelliCAL™
Supplier: Hach