ContactUsLinkComponent
Brother PT-P710BT Cube Plus Labeling Machine Prints durable labels up to 24 mm wide. Works with your PC, Mac, smartphone or tablet. USB and Bluetooth connectivity. Charges via your PC's USB port or a USB charger (adapter). Compact and mobile. 1 * 1 ST
Catalog # BROTPT-P710BT
Supplier:  BROTHER
Brother PT-P710BT Cube Plus Labeling Machine Prints durable labels up to 24 mm wide. Works with your PC, Mac, smartphone or tablet. USB and Bluetooth connectivity. Charges via your PC's USB port or a USB charger (adapter). Compact and mobile. 1 * 1 ST
Catalog # BROTPT-P710BT
Supplier:  BROTHER
Frequently Bought Together