Equipped with a 375 nm UV light and a switchable white light, the EdgePLUS AM4517MT-FUW high sensitivity digital microscope enhances UV features of paintings, trace evidence, and security print documents. 1 * 1 items
Catalog # IDCPAM4517MT-FUW
Supplier: Dino-Lite / IDCP B.V.
Frequently Bought Together