ContactUsLinkComponent
Homogenizer HS-30E, 200-5000 rpm, analog internal controller, as a set Set includes : Unit : HS-30E Clamp : CL200 for stirrer and homogeniser Plate stand with rod : ST110 plate 20x35 cm, rod dia. 16x h800 mm NOTE: No tissue grinder included! 1 * 1 items
Catalog # WITGDH.WOS01011
Supplier:  witeg Labortechnik
Homogenizer HS-30E, 200-5000 rpm, analog internal controller, as a set Set includes : Unit : HS-30E Clamp : CL200 for stirrer and homogeniser Plate stand with rod : ST110 plate 20x35 cm, rod dia. 16x h800 mm NOTE: No tissue grinder included! 1 * 1 items
Catalog # WITGDH.WOS01011
Supplier:  witeg Labortechnik
Frequently Bought Together