5-CHLOROPYRIDAZIN-3-AMINE 1 * 100 g
PHBLPB05523-100G
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
p-Benzoquinone, Supelco®
: Merck
Analytical:48-Well Custom Flat Bottom Base Plate for 12 x 32 Vials 1 * 1 ST
: MARKET SOURCE PART PROCESS
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
: Honeywell Chemicals
Xylene (mixture of isomers) ≥98.5% (isomers plus ethylbenzene) ACS
: Thermo Fisher Scientific
1,4-Dioxane, anhydrous ≥99.8%
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Formamide 99.5+%, extra pure
: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%
: Thermo Fisher Scientific