Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Katalognr. SARBMCA36S3CE24I
Anbieter: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Katalognr. SARBMCA36S3CE24I
Anbieter: Sartorius Balances
Frequently Bought Together
Gas cartridges, Campingaz®
Anbieter: CAMPING GAZ
Stainless steel sieves for ZM 200
Anbieter: Retsch GmbH
Magnetic hotplate stirrers, ARE/AREX 5 series
Anbieter: VELP SCIENTIFIC
Magnetic hotplate stirrers, C-MAG HS series
Anbieter: IKA
Vortex mixers, lab dancer
Anbieter: IKA
Magnetic stirring bars sets, IKAFLON®
Anbieter: IKA
Drying ovens with forced convection, FD series
Anbieter: Binder
Laboratory mixers
Anbieter: WARING BLENDOR
Accessories for mixer mills MM 200 and MM 400
Anbieter: Retsch GmbH
VWR®, Accessories for Analogue, Digital and Pulsing Vortex Shakers
Anbieter: VWR Collection
DrySyn® heating blocks
Anbieter: Asynt
VWR®, Accessories for Microplate Shakers and Orbital/ Mini Shakers
Anbieter: VWR Collection
Accessories for Memmert UN/UNplus and UF/UFplus ovens
Anbieter: MEMMERT
Accessories for knife mill, MICROTRON® MB 550
Anbieter: KINEMATICA
Globe stirrer couplings, BOLA
Anbieter: Bohlender
Ultrasonic liquid processors, programmable, Q500 and Q700
Anbieter: QSONICA LLC