ContactUsLinkComponent
SpectraMax PARADIGM Multimode Microplate Plate Reader Base Instrument. Includes: SoftMax Pro Software Standard 2 PMT Integrated S haking & Temperature Control Power Cable RS232 Serial Cable Compatible with 6-, 24- 96-, 384- & 1536-well plates 1 year 1 * 1 items
Catalog # MLDVPARADIGM-TR
Supplier:  Molecular Devices
SpectraMax PARADIGM Multimode Microplate Plate Reader Base Instrument. Includes: SoftMax Pro Software Standard 2 PMT Integrated S haking & Temperature Control Power Cable RS232 Serial Cable Compatible with 6-, 24- 96-, 384- & 1536-well plates 1 year 1 * 1 items
Catalog # MLDVPARADIGM-TR
Supplier:  Molecular Devices
Frequently Bought Together