ContactUsLinkComponent
SpectraMax iD3 Multi-Mode Microplate Detection Platform. System Includes: Spectral Absorbance, Fluorescence, and Luminescence detect ion, On-board Touchscreen for full acquisition, NFC Personalization, SoftMax Pro Software Integrated Shaking and Tem 1 * 1 items
Catalog # MLDVID3-AQ2KBNDLTR
Supplier:  Molecular Devices
SpectraMax iD3 Multi-Mode Microplate Detection Platform. System Includes: Spectral Absorbance, Fluorescence, and Luminescence detect ion, On-board Touchscreen for full acquisition, NFC Personalization, SoftMax Pro Software Integrated Shaking and Tem 1 * 1 items
Catalog # MLDVID3-AQ2KBNDLTR
Supplier:  Molecular Devices
Frequently Bought Together