ContactUsLinkComponent
SpectraMax ABS Single-Mode Microplate Reader. System Includes: Tunable Monochromator-based Spectral Absorbance (340nm to 850nm), Sof tMax Pro Software (4 perpetual licenses) Integrated Linear Shaking and Temperature Control from ambient + 5C to 45C, 1 * 1 items
Catalog # MLDVABS-TR
Supplier:  Molecular Devices
SpectraMax ABS Single-Mode Microplate Reader. System Includes: Tunable Monochromator-based Spectral Absorbance (340nm to 850nm), Sof tMax Pro Software (4 perpetual licenses) Integrated Linear Shaking and Temperature Control from ambient + 5C to 45C, 1 * 1 items
Catalog # MLDVABS-TR
Supplier:  Molecular Devices
Frequently Bought Together