ContactUsLinkComponent
SpectraMax PARADIGM CUSTOM, MULTI, type 680 Detection Cartridge offers 5 labels in FI, TRF, non-FRET and glow LUM extends the capabi lities of the detection system to work your specific assay. 1 * 1 items
Catalog # MLDV0200-7022-TR
Supplier:  Molecular Devices
SpectraMax PARADIGM CUSTOM, MULTI, type 680 Detection Cartridge offers 5 labels in FI, TRF, non-FRET and glow LUM extends the capabi lities of the detection system to work your specific assay. 1 * 1 items
Catalog # MLDV0200-7022-TR
Supplier:  Molecular Devices
Frequently Bought Together