ContactUsLinkComponent
SpectraMax AlphaScreen 384 STD Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen AlphaLisa assays. Compatible with SpectraMax PARADIGM and SpectraMax i3x systems. In addition, a patent pending desi 1 * 1 items
Catalog # MLDV0200-7017-TR
Supplier:  Molecular Devices
SpectraMax AlphaScreen 384 STD Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen AlphaLisa assays. Compatible with SpectraMax PARADIGM and SpectraMax i3x systems. In addition, a patent pending desi 1 * 1 items
Catalog # MLDV0200-7017-TR
Supplier:  Molecular Devices
Frequently Bought Together