ContactUsLinkComponent
SpectraMax Glow Luminescence (LUM) (96) Detection Cartridge for 96-well plates, utilizes a unique, well-isolating design that signif icantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax PARADIGM and/or SpectraMa 1 * 1 items
Catalog # MLDV0200-7014-TR
Supplier:  Molecular Devices
SpectraMax Glow Luminescence (LUM) (96) Detection Cartridge for 96-well plates, utilizes a unique, well-isolating design that signif icantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax PARADIGM and/or SpectraMa 1 * 1 items
Catalog # MLDV0200-7014-TR
Supplier:  Molecular Devices
Frequently Bought Together