ContactUsLinkComponent
SpectraMax Time Resolved Fluorescence (TRF) Detection Cartridge is built to measure Europium and Samarium, and is ideal for protein- protein interactions, GPCR and enzyme activity applications. Compatible with SpectraMax PARADIGM and/or SpectraMax i3 1 * 1 items
Catalog # MLDV0200-7008-TR
Supplier:  Molecular Devices
SpectraMax Time Resolved Fluorescence (TRF) Detection Cartridge is built to measure Europium and Samarium, and is ideal for protein- protein interactions, GPCR and enzyme activity applications. Compatible with SpectraMax PARADIGM and/or SpectraMax i3 1 * 1 items
Catalog # MLDV0200-7008-TR
Supplier:  Molecular Devices
Frequently Bought Together