ContactUsLinkComponent
SpectraMax Fluorescence Intensity (FI) (Cy3-Cy5) Detection Cartridge provides speed, sensitivity and flexibility. Compatible with S pectraMax PARADIGM and/or SpectraMax i3 systems. Optimized for Cy3-Cy5 fluorophores, the wavelength-tuned excitation l 1 * 1 items
Catalog # MLDV0200-7004-TR
Supplier:  Molecular Devices
SpectraMax Fluorescence Intensity (FI) (Cy3-Cy5) Detection Cartridge provides speed, sensitivity and flexibility. Compatible with S pectraMax PARADIGM and/or SpectraMax i3 systems. Optimized for Cy3-Cy5 fluorophores, the wavelength-tuned excitation l 1 * 1 items
Catalog # MLDV0200-7004-TR
Supplier:  Molecular Devices
Frequently Bought Together