Hydrocortisone Impurity F Pharmaceutical Secondary Standard Certified Reference Material 1 * 100 mg
Catalog # PHR2158-100MG
Anbieter: Merck
Hydrocortisone Impurity F Pharmaceutical Secondary Standard Certified Reference Material 1 * 100 mg
Catalog # PHR2158-100MG
Anbieter: Merck
Frequently Bought Together
VWR®, Conductivity standard solutions
Anbieter: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Anbieter: VWR Chemicals
Buffer solutions in sachets
Anbieter: Mettler - Toledo
Accessories for chloride analyser, model 926
Anbieter: SHERWOOD SCIENTIFIC INSTRUMENT
Buffer solutions in sachets, Certipur®, Supelco®
Anbieter: Merck
UV/Visible standards, Supelco®
Anbieter: Merck
pH buffer solutions
Anbieter: Mettler - Toledo
Conductivity standard 1,413 µS/cm (25°C; 0,01 mol/L; KCl) for conductivity measurement
Anbieter: SI Analytics
Turbidity standards, Stablcal®
Anbieter: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Anbieter: Lovibond