Hydrocortisone Impurity E Pharmaceutical Secondary Standard Certified Reference Material 1 * 100 mg
Catalog # PHR2157-100MG
Supplier: Merck
Hydrocortisone Impurity E Pharmaceutical Secondary Standard Certified Reference Material 1 * 100 mg
Catalog # PHR2157-100MG
Supplier: Merck
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffer solutions
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer solutions, NIST/DIN
Supplier: Mettler - Toledo