OXIVIR PLUS NC SPRAY 6X0.75L 1 * 6 Fl
Catalog # DIVY101106359
Supplier: Diversey
Frequently Bought Together
Petroleum spirit, 40…60 °C, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute, SupraSolv® for GC-ECD/FID
Supplier: Merck
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Absorbent powder, Chemizorb® Alkalis
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck