ContactUsLinkComponent
,,9 components: Al 50mg/l Cu 50mg/l Fe 50mg/l Na 50mg/l Ni 50mg/l Mg 50mg/l K 50mg/l Ca 1000mg/l Si 500mg/l in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM096634L1
,,9 components: Al 50mg/l Cu 50mg/l Fe 50mg/l Na 50mg/l Ni 50mg/l Mg 50mg/l K 50mg/l Ca 1000mg/l Si 500mg/l in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM096634L1
Frequently Bought Together