L-MARK Gold Syringe 100UL Fixed Needle, PTFE, 51mm Length, 22s Gauge, LC Tip PK2 1 * 2 items
Catalog # SGEALMK.2620719C2P
Supplier: Trajan Scientific and Medical
L-MARK Gold Syringe 100UL Fixed Needle, PTFE, 51mm Length, 22s Gauge, LC Tip PK2 1 * 2 items
Catalog # SGEALMK.2620719C2P
Supplier: Trajan Scientific and Medical
Frequently Bought Together
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Spillage trays
Supplier: S.C.A.T.
VWR® Air Valve
Supplier: VWR Collection
HPLC columns, LiChrospher®
Supplier: Merck
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Vydac 214TP C4, HPLC columns
Supplier: Avantor
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
PHASE SEPARATOR 150ML (25/PK) 1 * 25 items
Supplier: BIOTAGE