Auftragserfassung
Switzerland
ContactUsLinkComponent
EPQpYEEIPIYL, a phosphopeptide, is a Src homology 2 (SH2) domain ligand. EPQpYEEIPIYL activates Src family members (e.g. Lck, Hck, F yn) by binding to SH2 domains. 1 * 1 mg
Catalog # MDEXHY-P3279-1MG
Supplier:  MEDCHEMEXPRESS
EPQpYEEIPIYL, a phosphopeptide, is a Src homology 2 (SH2) domain ligand. EPQpYEEIPIYL activates Src family members (e.g. Lck, Hck, F yn) by binding to SH2 domains. 1 * 1 mg
Catalog # MDEXHY-P3279-1MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together