5-chloropyridazin-3-amine,97% 1 * 10 g
PHBLPB05523-10G
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals