4-chloro-5H,6H,7H-pyrrolo[3,4-d]pyrimidine hydrochloride 1 * 1 g
N° de catalogue PHBLPBZX1493-1-1G
Fournisseur: Pharmablock (USA)
4-chloro-5H,6H,7H-pyrrolo[3,4-d]pyrimidine hydrochloride 1 * 1 g
N° de catalogue PHBLPBZX1493-1-1G
Fournisseur: Pharmablock (USA)
Frequently Bought Together
Tetrahydrofuran, anhydrous (max. 0.003% H₂O) ≥99.9% stabilised
Fournisseur: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
Fournisseur: VWR Chemicals
Acetonitrile ≥99.95%, HiPerSolv CHROMANORM® ULTRA for LC-MS, suitable for UPLC/UHPLC-MS instruments
Fournisseur: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Fournisseur: VWR Chemicals
Toluene ≥98%, TECHNICAL
Fournisseur: VWR Chemicals
N,N-Dimethylacetamide 99.99-100.00%, PESTINORM® for headspace gas chromatography
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Fournisseur: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Fournisseur: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Fournisseur: VWR Chemicals