1-(5-bromopyrimidin-2-yl)cyclopropane-1-carboxylic acid 1 * 1 g
Catalog # PHBLPBZX1489-1G
Supplier: Pharmablock (USA)
1-(5-bromopyrimidin-2-yl)cyclopropane-1-carboxylic acid 1 * 1 g
Catalog # PHBLPBZX1489-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Sulphuryl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Dimethyl sulphoxide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
n-Hexane 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™
Supplier: Honeywell Chemicals