3,3-dimethyloxan-4-ol 1 * 1 g
PHBLPBZ0201-1G
Frequently Bought Together
2-Propanol ≥99.8%, SPECTRONORM® for spectroscopy
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Diethylene glycol monobutyl ether ≥99%
: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS
: Thermo Fisher Scientific