3-(trifluoromethoxy)pyridine-2-carboxylic acid 1 * 1 g
PHBLPBYS1715-1G
Frequently Bought Together
n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals