(2S,4R)-1-[(2S)-2-(11-aminoundecanoylamino)-3,3-dimethyl-butanoyl]-4-hydroxy-N-[(1S)-1-[4-(4-methylthiazol-5-yl)phenyl]ethyl]pyrroli dine-2-carboxamide 1 * 1 g
Catalog # PHBLPBY2074-1G
Supplier: Pharmablock (USA)
(2S,4R)-1-[(2S)-2-(11-aminoundecanoylamino)-3,3-dimethyl-butanoyl]-4-hydroxy-N-[(1S)-1-[4-(4-methylthiazol-5-yl)phenyl]ethyl]pyrroli dine-2-carboxamide 1 * 1 g
Catalog # PHBLPBY2074-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals