(2,5-dioxopyrrolidin-1-yl) 3-[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxo-propoxy]propanoate 1 * 1 g
Catalog # PHBLPBY0753-1G
Supplier: Pharmablock (USA)
(2,5-dioxopyrrolidin-1-yl) 3-[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxo-propoxy]propanoate 1 * 1 g
Catalog # PHBLPBY0753-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-2-Methyltetrahydrofuran, EMPLURA®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Phenetole ≥98%
Supplier: Thermo Fisher Scientific
Methanol ≥99.7% (by GC), Puriss., meets analytical specification of Ph.Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
Formamide ≥99%
Supplier: Thermo Fisher Scientific
n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.9%
Supplier: Honeywell Chemicals