tert-butyl 3-[tert-butyl(dimethyl)silyl]oxy-6-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]pyrrole-4-carboxylate 1 * 1 g
Catalog # PHBLPBWZ267-1G
Supplier: Pharmablock (USA)
tert-butyl 3-[tert-butyl(dimethyl)silyl]oxy-6-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]pyrrole-4-carboxylate 1 * 1 g
Catalog # PHBLPBWZ267-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals